Learn More
Apexbio Technology LLC 3-Methylthiopropionic-acid
SDP
Catalog No. 5000569955
Encompass_Preferred
3-(Methylthio)propionic acid (CAS No 646-01-5) is a compound commonly used in biochemical and pharmacological research It is known for its role in metabolic studies often investigated for its effects on thiol pathways and methionine metabolism Its mechanism of action involves interactions with sulfur-containing amino acids influencing enzymatic processes and redox balance in cells In vitro the compound exhibits activity typically observed in nanomolar to low micromolar ranges making it suitable for precise modulation studies It is frequently applied in various cell models and animal studies to explore its potential therapeutic implications and as a candidate for drug screening Experimental concentrations typically depend on the specific research design allowing for tailored investigation into its physiological effects and biomedical applications
Catalog No. 50-005-69955 Supplier Apexbio Technology LLC Supplier No. C7197200
Add to Cart
Add to Cart
3-(Methylthio)propionic acid (CAS No 646-01-5) is a compound commonly used in biochemical and pharmacological research It is known for its role in metabolic studies often investigated for its effects on thiol pathways and methionine metabolism Its mechanism of action involves interactions with sulfur-containing amino acids influencing enzymatic processes and redox balance in cells In vitro the compound exhibits activity typically observed in nanomolar to low micromolar ranges making it suitable for precise modulation studies It is frequently applied in various cell models and animal studies to explore its potential therapeutic implications and as a candidate for drug screening Experimental concentrations typically depend on the specific research design allowing for tailored investigation into its physiological effects and biomedical applications