Learn More
Medchemexpress LLC 3-methyltoxoflavin | 32502-62-8 | 98.6% | 207.19 g·mol-1 | C8H9N5O2 | 5 MG
SDP
Catalog No. 501968824
Encompass_Preferred
3-methyltoxoflavin is a small-molecule research compound that potently inhibits protein disulfide isomerase (PDI) with a reported IC50 of 170 nM. Supplied as a high-purity powder for biochemical and cell-based studies, it is intended for research use only.
  • Potent protein disulfide isomerase (PDI) inhibitor with IC50 = 170 nM.
  • High purity: 98.62%.
  • Molecular weight: 207.19 g·mol-1; chemical formula C8H9N5O2.
  • Available in multiple quantities and supplied as a dry powder.
  • Powder storage stability: -20°C for 3 years, 4°C for 2 years.
Catalog No. 50-196-8824 Supplier Medchemexpress LLC Supplier No. HY1111175MG
3-methyltoxoflavin is a small-molecule research compound that potently inhibits protein disulfide isomerase (PDI) with a reported IC50 of 170 nM. Supplied as a high-purity powder for biochemical and cell-based studies, it is intended for research use only.
  • Potent protein disulfide isomerase (PDI) inhibitor with IC50 = 170 nM.
  • High purity: 98.62%.
  • Molecular weight: 207.19 g·mol-1; chemical formula C8H9N5O2.
  • Available in multiple quantities and supplied as a dry powder.
  • Powder storage stability: -20°C for 3 years, 4°C for 2 years.