Learn More
TARGETMOL CHEMICALS INC 3-Methyluridine 10MG
SDP
Catalog No. 502876974
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.3-Methyluridine is a modified RNA nucleoside of cellular RNA and commonly found in the form of RNA modifications. Purity 99.14%
Catalog No. 50-287-6974 Supplier TARGETMOL CHEMICALS INC Supplier No. T1349910MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.3-Methyluridine is a modified RNA nucleoside of cellular RNA and commonly found in the form of RNA modifications. Purity 99.14%