Learn More
Medchemexpress LLC 3-Methyluridine | 2140-69-4 | 99.05% | 258.23 | 1 ML
SDP
Catalog No. 5000437332
Encompass_Preferred
3-Methyluridine (m3U) is a methylated nucleotide found in ribosomal RNA (rRNA), primarily targeting specific base sites like 23S rRNA. It introduces a methyl group at the N3 position of uracil, influencing RNA's secondary structure stability and base pairing. This regulation impacts ribosome function, including ribosomal subunit binding and tRNA interaction.
  • Utilized as a key raw material for synthesizing modified nucleotides.
  • Constructs RNA oligonucleotides with methylation modifications.
  • Used in studying the effects of RNA methylation on gene expression and drug resistance.
Catalog No. 50-004-37332 Supplier Medchemexpress LLC Supplier No. HY11313810MM/1ML
3-Methyluridine (m3U) is a methylated nucleotide found in ribosomal RNA (rRNA), primarily targeting specific base sites like 23S rRNA. It introduces a methyl group at the N3 position of uracil, influencing RNA's secondary structure stability and base pairing. This regulation impacts ribosome function, including ribosomal subunit binding and tRNA interaction.
  • Utilized as a key raw material for synthesizing modified nucleotides.
  • Constructs RNA oligonucleotides with methylation modifications.
  • Used in studying the effects of RNA methylation on gene expression and drug resistance.