Learn More
Medchemexpress LLC 3-Methyluridine | 2140-69-4 | 99.1% | 258.23 | 50 MG
SDP
Catalog No. 5000435774
Encompass_Preferred
3-Methyluridine is a methylated nucleotide found in ribosomal RNA. It introduces a methyl group at the N3 position of uracil, influencing the secondary structure stability and base pairing ability of RNA, and regulating ribosome function.
  • Primarily targets specific base sites of RNA molecules such as 23S rRNA.
  • Regulates ribosome function, including ribosomal subunit binding and tRNA interaction.
  • Utilized as a key raw material for synthesizing modified nucleotides.
  • Used for constructing RNA oligonucleotides with methylation modifications.
  • Helps study the effects of RNA methylation on gene expression and drug resistance.
Catalog No. 50-004-35774 Supplier Medchemexpress LLC Supplier No. HY11313850MG
3-Methyluridine is a methylated nucleotide found in ribosomal RNA. It introduces a methyl group at the N3 position of uracil, influencing the secondary structure stability and base pairing ability of RNA, and regulating ribosome function.
  • Primarily targets specific base sites of RNA molecules such as 23S rRNA.
  • Regulates ribosome function, including ribosomal subunit binding and tRNA interaction.
  • Utilized as a key raw material for synthesizing modified nucleotides.
  • Used for constructing RNA oligonucleotides with methylation modifications.
  • Helps study the effects of RNA methylation on gene expression and drug resistance.