Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Methylvaleric Acid | 105-43-1 | 97.5% | 116.16 | 25 G
SDP

Catalog No. 5000451656
Encompass_Preferred

3-Methylvaleric Acid is a branched alkanoic acid that can be utilized by *Pseudomonas citronettotis* as a sole carbon source for its growth. Its derivatives exhibit anti-hyperalgesic and anticonvulsant activities.

  • Branched alkanoic acid
  • Can be utilized by *Pseudomonas citronettotis* as a sole carbon source
  • Derivatives show anti-hyperalgesic and anticonvulsant activities
  • For research use only
  • Appearance: liquid (density: 0.9262 g/cm³)
  • Color: colorless to light yellow
  • Initial source: endogenous metabolite

Catalog No. 50-004-51656 Supplier Medchemexpress LLC Supplier No. 25GHYW010513
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-Methylvaleric Acid is a branched alkanoic acid that can be utilized by *Pseudomonas citronettotis* as a sole carbon source for its growth. Its derivatives exhibit anti-hyperalgesic and anticonvulsant activities.

  • Branched alkanoic acid
  • Can be utilized by *Pseudomonas citronettotis* as a sole carbon source
  • Derivatives show anti-hyperalgesic and anticonvulsant activities
  • For research use only
  • Appearance: liquid (density: 0.9262 g/cm³)
  • Color: colorless to light yellow
  • Initial source: endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.