Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Methylvaleric acid | 105-43-1 | 99.99% | 116.16 | 10 G
SDP

Catalog No. 5000451800
Encompass_Preferred

3-Methylvaleric Acid is a branched alkanoic acid that can be utilized by *Pseudomonas citronettotis* as a sole carbon source for its growth. Derivatives of 3-Methylvaleric Acid have shown anti-hyperalgesic and anticonvulsant activities.

  • Purity: 99.99%
  • Appearance: Liquid (density: 0.9262 g/cm³)
  • Color: Colorless to light yellow
  • Initial source: Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-51800 Supplier Medchemexpress LLC Supplier No. HYW01051310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Methylvaleric Acid is a branched alkanoic acid that can be utilized by *Pseudomonas citronettotis* as a sole carbon source for its growth. Derivatives of 3-Methylvaleric Acid have shown anti-hyperalgesic and anticonvulsant activities.

  • Purity: 99.99%
  • Appearance: Liquid (density: 0.9262 g/cm³)
  • Color: Colorless to light yellow
  • Initial source: Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.