Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Methylvaleric Acid | 105-43-1 | MFCD00002729 | 99.99% | 116.16 | 1 ML
SDP

Catalog No. 5000451583
Encompass_Preferred

3-Methylvaleric Acid is a branched alkanoic acid utilized by Pseudomonas citronettotis as a sole carbon source. Its derivatives exhibit anti-hyperalgesic and anticonvulsant activities. This product is supplied as a 10 mM solution in 1 mL of DMSO.

  • Purity of 99.99%
  • Molecular weight of 116.16
  • Provided as a colorless to light yellow liquid
  • Recommended storage in solvent at -80°C for 6 months

Catalog No. 50-004-51583 Supplier Medchemexpress LLC Supplier No. HYW01051310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Methylvaleric Acid is a branched alkanoic acid utilized by Pseudomonas citronettotis as a sole carbon source. Its derivatives exhibit anti-hyperalgesic and anticonvulsant activities. This product is supplied as a 10 mM solution in 1 mL of DMSO.

  • Purity of 99.99%
  • Molecular weight of 116.16
  • Provided as a colorless to light yellow liquid
  • Recommended storage in solvent at -80°C for 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.