Learn More
Medchemexpress LLC 3-Methylvaleric acid (Standard) | 105-43-1 | 682090 | 99.5% | 116.16 | 50 MG
SDP
Catalog No. 5000366368
Encompass_Preferred
3-Methylvaleric acid (Standard) is an analytical standard form of 3-Methylvaleric acid (HY-W010513) intended for research and analytical applications. It is a branched alkanoic acid that can be utilized by *Pseudomonas citronettotis* as a sole carbon source for its growth. Its derivatives exhibit anti-hyperalgesic and anticonvulsant activities. This compound is of analytical standard grade, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard grade.
  • Can be utilized by *Pseudomonas citronettotis* as a sole carbon source.
  • Derivatives exhibit anti-hyperalgesic and anticonvulsant activities.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-66368 Supplier Medchemexpress LLC Supplier No. HYW010513R50MG
3-Methylvaleric acid (Standard) is an analytical standard form of 3-Methylvaleric acid (HY-W010513) intended for research and analytical applications. It is a branched alkanoic acid that can be utilized by *Pseudomonas citronettotis* as a sole carbon source for its growth. Its derivatives exhibit anti-hyperalgesic and anticonvulsant activities. This compound is of analytical standard grade, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard grade.
  • Can be utilized by *Pseudomonas citronettotis* as a sole carbon source.
  • Derivatives exhibit anti-hyperalgesic and anticonvulsant activities.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.