Learn More
Medchemexpress LLC 3-methylvaleric acid (standard) | 105-43-1 | 99.5% | 116.16 | 500 MG
SDP
Catalog No. 5000451749
Encompass_Preferred
3-Methylvaleric acid (Standard) is an analytical standard form of 3-Methylvaleric acid intended for research and analytical applications. It is a branched alkanoic acid that can be utilized by *Pseudomonas citronettotis* as a sole carbon source for its growth. Derivatives of 3-Methylvaleric acid exhibit anti-hyperalgesic and anticonvulsant activities.
  • Intended for research and analytical applications
  • Branched alkanoic acid
  • Utilized by *Pseudomonas citronettotis* as a sole carbon source
  • Derivatives exhibit anti-hyperalgesic and anticonvulsant activities
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
  • Suitable for use in HPLC, GC, and MS
  • Appears as a colorless to light yellow liquid
Catalog No. 50-004-51749 Supplier Medchemexpress LLC Supplier No. HYW010513R500MG
3-Methylvaleric acid (Standard) is an analytical standard form of 3-Methylvaleric acid intended for research and analytical applications. It is a branched alkanoic acid that can be utilized by *Pseudomonas citronettotis* as a sole carbon source for its growth. Derivatives of 3-Methylvaleric acid exhibit anti-hyperalgesic and anticonvulsant activities.
  • Intended for research and analytical applications
  • Branched alkanoic acid
  • Utilized by *Pseudomonas citronettotis* as a sole carbon source
  • Derivatives exhibit anti-hyperalgesic and anticonvulsant activities
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
  • Suitable for use in HPLC, GC, and MS
  • Appears as a colorless to light yellow liquid