Learn More
Medchemexpress LLC 3-Methylxanthine | 1076-22-8 | 99.45% | C6H6N4O2 | 25 G
SDP
Catalog No. 5000358645
Encompass_Preferred
3-Methylxanthine is a xanthine derivative and a cyclic guanosine monophosphate (GMP) inhibitor, with an IC50 of 920 μM on guinea-pig isolated trachealis muscle. It is an endogenous metabolite.
  • Xanthine derivative
  • Cyclic guanosine monophosphate (GMP) inhibitor
  • IC50 of 920 μM on guinea-pig isolated trachealis muscle
  • Endogenous metabolite
  • Available in solid and solution forms
Catalog No. 50-003-58645 Supplier Medchemexpress LLC Supplier No. 25GHY50723
3-Methylxanthine is a xanthine derivative and a cyclic guanosine monophosphate (GMP) inhibitor, with an IC50 of 920 μM on guinea-pig isolated trachealis muscle. It is an endogenous metabolite.
  • Xanthine derivative
  • Cyclic guanosine monophosphate (GMP) inhibitor
  • IC50 of 920 μM on guinea-pig isolated trachealis muscle
  • Endogenous metabolite
  • Available in solid and solution forms