Learn More
Medchemexpress LLC 3-Methylxanthine | 1076-22-8 | 99.5% | C6H6N4O2 | 1 ML
SDP
Catalog No. 5000359126
Encompass_Preferred
3-Methylxanthine is a xanthine derivative and a cyclic guanosine monophosphate (GMP) inhibitor, with an IC50 of 920 μM on guinea-pig isolated trachealis muscle. This product is provided as a solid with a purity of 99.45%.
  • Xanthine derivative
  • Cyclic guanosine monophosphate (GMP) inhibitor
  • IC50 of 920 μM on guinea-pig isolated trachealis muscle
  • Provided as a solid
  • Purity of 99.45% (HPLC)
  • Stable under recommended storage conditions
Catalog No. 50-003-59126 Supplier Medchemexpress LLC Supplier No. HY5072310MM/1ML
3-Methylxanthine is a xanthine derivative and a cyclic guanosine monophosphate (GMP) inhibitor, with an IC50 of 920 μM on guinea-pig isolated trachealis muscle. This product is provided as a solid with a purity of 99.45%.
  • Xanthine derivative
  • Cyclic guanosine monophosphate (GMP) inhibitor
  • IC50 of 920 μM on guinea-pig isolated trachealis muscle
  • Provided as a solid
  • Purity of 99.45% (HPLC)
  • Stable under recommended storage conditions