Learn More
Medchemexpress LLC 3-Methylxanthine | 1076-22-8 | 99.5% | C6H6N4O2 | 100 G
SDP
Catalog No. 5000358320
Encompass_Preferred
3-Methylxanthine is a xanthine derivative that acts as a cyclic guanosine monophosphate (GMP) inhibitor, with an IC50 of 920 μM on guinea-pig isolated trachealis muscle. This product is intended for research use only.
  • Stable under recommended storage conditions
  • Recommended storage for powder at -20°C for 3 years or 4°C for 2 years
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-58320 Supplier Medchemexpress LLC Supplier No. HY50723100G
3-Methylxanthine is a xanthine derivative that acts as a cyclic guanosine monophosphate (GMP) inhibitor, with an IC50 of 920 μM on guinea-pig isolated trachealis muscle. This product is intended for research use only.
  • Stable under recommended storage conditions
  • Recommended storage for powder at -20°C for 3 years or 4°C for 2 years
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month