Learn More
Medchemexpress LLC 3-Methylxanthine | 1076-22-8 | 99.5% | C6H6N4O2 | 25 MG
SDP
Catalog No. 5000358899
Encompass_Preferred
3-Methylxanthine is a xanthine derivative and an endogenous metabolite, primarily known as a cyclic guanosine monophosphate (GMP) inhibitor. It is intended for research use only.
  • Acts as a cyclic GMP inhibitor
  • IC50 of 920 μM on guinea-pig isolated trachealis muscle
  • Classified as an endogenous metabolite
  • Solid appearance, white to khaki in color
  • Molecular structure consistent with NMR and LCMS analysis
Catalog No. 50-003-58899 Supplier Medchemexpress LLC Supplier No. 25MGHY50723R
3-Methylxanthine is a xanthine derivative and an endogenous metabolite, primarily known as a cyclic guanosine monophosphate (GMP) inhibitor. It is intended for research use only.
  • Acts as a cyclic GMP inhibitor
  • IC50 of 920 μM on guinea-pig isolated trachealis muscle
  • Classified as an endogenous metabolite
  • Solid appearance, white to khaki in color
  • Molecular structure consistent with NMR and LCMS analysis