Learn More
Medchemexpress LLC 3-Methylxanthine | 1076-22-8 | MFCD00005580 | 99.5% | C6H6N4O2 | 50 G
SDP
Catalog No. 5000358062
Encompass_Preferred
3-Methylxanthine is a xanthine derivative that acts as a cyclic guanosine monophosphate (GMP) inhibitor, exhibiting an IC50 of 920 μM on guinea-pig isolated trachealis muscle. It is classified as an endogenous metabolite.
  • Xanthine derivative
  • Cyclic guanosine monophosphate (GMP) inhibitor
  • Exhibits an IC50 of 920 μM on guinea-pig isolated trachealis muscle
  • Endogenous metabolite
  • Used for research purposes only
Catalog No. 50-003-58062 Supplier Medchemexpress LLC Supplier No. HY5072350G
3-Methylxanthine is a xanthine derivative that acts as a cyclic guanosine monophosphate (GMP) inhibitor, exhibiting an IC50 of 920 μM on guinea-pig isolated trachealis muscle. It is classified as an endogenous metabolite.
  • Xanthine derivative
  • Cyclic guanosine monophosphate (GMP) inhibitor
  • Exhibits an IC50 of 920 μM on guinea-pig isolated trachealis muscle
  • Endogenous metabolite
  • Used for research purposes only