Learn More
Selleck Chemical LLC 3-Methylxanthine
SDP
Catalog No. 5000003297
Encompass_Preferred
3-Methylxanthine (3-MX) a 3-alkylxanthine is an inhibitor of cyclic guanosine monophosphate (GMP) phosphodiesterase (PDE) with IC50 of 920 M on guinea-pig isolated trachealis muscle
Catalog No. 50-000-03297 Supplier Selleck Chemical LLC Supplier No. S6186100MG
3-Methylxanthine (3-MX) a 3-alkylxanthine is an inhibitor of cyclic guanosine monophosphate (GMP) phosphodiesterase (PDE) with IC50 of 920 M on guinea-pig isolated trachealis muscle