Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylxanthine, 99.43%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15168680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15168680 100 mg
ICN15168683 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15168680 Supplier MP Biomedicals, Inc Supplier No. 0215168680
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1076-22-8
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
InChI Key GMSNIKWWOQHZGF-UHFFFAOYSA-N
Synonym 3-methylxanthine, 3-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-3-methylpurine, xanthine, 3-methyl, 3 mx, 3-methyl xanthine, 1h-purine-2,6-dione, 3,7-dihydro-3-methyl, unii-ws6x982oec, 3-methyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione, ccris 5817
PubChem CID 70639
ChEBI CHEBI:62207
IUPAC Name 3-methyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)NC1=O)NC=N2

Specifications

Color White
Quantity 100 mg
Formula Weight 166.1
Physical Form Powder
Chemical Name or Material 3-Methylxanthine, For HPLC analysis
EINECSNumber 214-058-1
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.