Learn More
Medchemexpress LLC 3-methylxanthine (standard) | 1076-22-8 | 99.6% | C6H6N4O2 | 50 MG
SDP
Catalog No. 5000357793
Encompass_Preferred
3-Methylxanthine (Standard) serves as the analytical standard for 3-Methylxanthine, a xanthine derivative and cyclic guanosine monophosphate (GMP) inhibitor. Intended for research and analytical applications, this standard has an IC50 of 920 μM on guinea-pig isolated trachealis muscle.
  • Used as an analytical reference standard in various assays.
  • Suitable for qualitative, quantitative, and methodological research.
  • Applicable in techniques such as HPLC, GC, and MS.
  • Intended for research applications only.
Catalog No. 50-003-57793 Supplier Medchemexpress LLC Supplier No. HY50723R50MG
3-Methylxanthine (Standard) serves as the analytical standard for 3-Methylxanthine, a xanthine derivative and cyclic guanosine monophosphate (GMP) inhibitor. Intended for research and analytical applications, this standard has an IC50 of 920 μM on guinea-pig isolated trachealis muscle.
  • Used as an analytical reference standard in various assays.
  • Suitable for qualitative, quantitative, and methodological research.
  • Applicable in techniques such as HPLC, GC, and MS.
  • Intended for research applications only.