Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ 3 mL PBS in 10 mL Sterile Tubes
SDP

Catalog No. 21520183
Change view
Click to view available options
Quantity:
750/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-183 750/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-520-183 Supplier Cenmed™ Supplier No. 38500004QT
Add to Cart
Add to Cart
This item is not returnable. View return policy

10 mL sterile tubes filled with 3 mL PBS (Phosphate Buffered Saline).

  • 3 mL filled PBS (Phosphate-Buffer-Saline)
  • 10 mL Tubes
  • Sterile Tubes
  • Dulbecco
  • Made in the USA

Specifications

Sterility Sterile
Quantity 750/Pk.
Volume (Metric) 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.