Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ 3 mL VTM in 10 mL Sterile Tubes
SDP

Catalog No. 21520184
Change view
Click to view available options
Quantity:
750/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-184 750/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-520-184 Supplier Cenmed™ Supplier No. C38900100QT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

10 mL sterile tubes filled with 3 mL VTM (Viral Transport Medium).

  • Suitable for collection, transport, maintenance and storage of clinical specimens containing viruses
  • Room Temperature Stability
  • 3 mL filled VTM (Viral Transport Medium)
  • 10 mL Tubes
  • Made in the USA

Specifications

Sterility Sterile
Quantity 750/Pk.
Volume (Metric) 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.