Learn More
Chemglass Life Sciences Beads, Borosilicate Glass, 3mm, Approx. 12, 500 Pcs. Per Lb.
SDP
Catalog No. CG110102
Encompass_Preferred
Beads, Borosilicate Glass, 3mm, Approx. 12,500 Pcs. Per Lb. Solid borosilicate spheres can be used as distillation column packing, boiling stones or to provide support for chromatography columns used in gel filtrations. Beads have excellent chemical resistance. Packed in boxes containing one pound.
Catalog No. CG110102 Supplier Chemglass Life Sciences Supplier No. CG110102
Beads, Borosilicate Glass, 3mm, Approx. 12,500 Pcs. Per Lb. Solid borosilicate spheres can be used as distillation column packing, boiling stones or to provide support for chromatography columns used in gel filtrations. Beads have excellent chemical resistance. Packed in boxes containing one pound.