Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Morpholinecarboxamide, N-[4-[[5-(1,6-dimethyl-1H-pyrazolo[3,4-b]pyridin-4-yl)-... | 2205095-75-4 | 99.9% | 532.68 g/mol | C29H40N8O2 | 5 MG
SDP

Catalog No. 5000221413
Encompass_Preferred

TLR7/8-IN-1 is a crystalline form of a dual TLR7/TLR8 inhibitor (patent WO2019220390, compound 2b) provided for laboratory research, including studies of autoimmune disease. The compound is supplied as a white to off-white solid or as DMSO solutions, and is characterized by defined molecular and purity data for reproducible preclinical use.

  • High purity: 99.92%.
  • Molecular weight: 532.68 g/mol.
  • Chemical formula: C29H40N8O2.
  • Soluble in DMSO (12.5 mg/mL; ultrasonic recommended).
  • Available as milligram quantities or 10 mM DMSO solutions for experimental use.
  • Storage stability: powder -20 °C (3 years); in solvent -80 °C (6 months).

Catalog No. 50-002-21413 Supplier Medchemexpress LLC Supplier No. HY1393235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TLR7/8-IN-1 is a crystalline form of a dual TLR7/TLR8 inhibitor (patent WO2019220390, compound 2b) provided for laboratory research, including studies of autoimmune disease. The compound is supplied as a white to off-white solid or as DMSO solutions, and is characterized by defined molecular and purity data for reproducible preclinical use.

  • High purity: 99.92%.
  • Molecular weight: 532.68 g/mol.
  • Chemical formula: C29H40N8O2.
  • Soluble in DMSO (12.5 mg/mL; ultrasonic recommended).
  • Available as milligram quantities or 10 mM DMSO solutions for experimental use.
  • Storage stability: powder -20 °C (3 years); in solvent -80 °C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.