Learn More
Medchemexpress LLC 3-morpholino-2-hydroxypropanesulfonic acid | 68399-77-9 | MFCD00009629 | 99.7% | 225.26 g/mol | C7H15NO5S | 500 G
SDP
Catalog No. 5000248025
Encompass_Preferred
MOPSO is a zwitterionic biological buffer used to maintain stable near-neutral pH in biochemical and cell culture applications. It buffers effectively in the 6.2-7.6 pH range and is supplied as a high-purity powder with supporting analytical documentation for quality control.
  • Useful pH range of 6.2-7.6.
  • High purity suitable for research applications.
  • Appropriate for biological and biochemical buffering needs.
  • Supplied as a powder for convenient storage and handling.
  • Analytical documentation available (COA, SDS, HNMR, RP-HPLC, MS).
Catalog No. 50-002-48025 Supplier Medchemexpress LLC Supplier No. HY101850500G
Add to Cart
Add to Cart
MOPSO is a zwitterionic biological buffer used to maintain stable near-neutral pH in biochemical and cell culture applications. It buffers effectively in the 6.2-7.6 pH range and is supplied as a high-purity powder with supporting analytical documentation for quality control.
  • Useful pH range of 6.2-7.6.
  • High purity suitable for research applications.
  • Appropriate for biological and biochemical buffering needs.
  • Supplied as a powder for convenient storage and handling.
  • Analytical documentation available (COA, SDS, HNMR, RP-HPLC, MS).