Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-morpholinobenzaldehyde, 97%, Thermo Scientific™

Catalog No. CC33804DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC33804DE 5 g Amber Glass Bottle
CC33804CB 250 mg Amber Glass Bottle
CC33804DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC33804DE Supplier Thermo Scientific Chemicals Supplier No. CC33804DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 446866-87-1
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
InChI Key LQORKFSMUOSSQM-UHFFFAOYSA-N
Synonym 3-morpholinobenzaldehyde, 3-morpholin-4-yl benzaldehyde, 4-3-formylphenyl morpholine, 3-4-morpholinyl benzaldehyde, 3-morpholin-4-yl-benzaldehyde, pubchem13334, 3-morpholin-4-yl benzaldehyd, benzaldehyde,3-4-morpholinyl
PubChem CID 7164584
IUPAC Name 3-morpholin-4-ylbenzaldehyde
SMILES C1COCCN1C2=CC=CC(=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 191.23
Percent Purity 97%
Chemical Name or Material 3-Morpholinobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.