Learn More
Abcam 3-Morpholinosydnonimine hydrochloride (SIN-1 hydrochloride), Vasodilator, 20MG
Catalog No. 502658129
Encompass_Preferred
MW 206.63 Da, Purity >99%. Releases NO, superoxide and peroxynitrite under physiological conditions. Potent vasodilator and inhibitor of platelet aggregation. Active metabolite of molsidomine (ab142914). Protective in myocardial ischemia-reperfusion. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-8129 Supplier Abcam Supplier No. AB14152520MG
MW 206.63 Da, Purity >99%. Releases NO, superoxide and peroxynitrite under physiological conditions. Potent vasodilator and inhibitor of platelet aggregation. Active metabolite of molsidomine (ab142914). Protective in myocardial ischemia-reperfusion. Orally active.
The product is subject to the following: Abcam Restricted Use Statement