Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC 3-(N-Fmoc-amino)piperidine-3-carboxylic acid HCl | 5 g | CAS 368866-20-0 | MDL MFCD04113564

Catalog No. 502350192
Encompass

3-(N-Fmoc-amino)piperidine-3-carboxylic acid HCl is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 368866-20-0
- MDL: MFCD04113564
- InChIKey: GZXZDFOJUNNXPX-UHFFFAOYSA-N
- Molecular Weight: 402.875
- Molecular Formula: C21H23ClN2O4
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29333999
- Country Of Origin: China
- IUPAC: 3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)piperidine-3-carboxylic acid hydrochloride
- SMILES: O=C(OCC1C2=CC=CC=C2C3=C1C=CC=C3)NC4(CCCNC4)C(O)=O.Cl

Catalog No. 50-235-0192 Supplier STA PHARMACEUTICAL US LLC Supplier No. STAAA102055GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-(N-Fmoc-amino)piperidine-3-carboxylic acid HCl is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 368866-20-0
- MDL: MFCD04113564
- InChIKey: GZXZDFOJUNNXPX-UHFFFAOYSA-N
- Molecular Weight: 402.875
- Molecular Formula: C21H23ClN2O4
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29333999
- Country Of Origin: China
- IUPAC: 3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)piperidine-3-carboxylic acid hydrochloride
- SMILES: O=C(OCC1C2=CC=CC=C2C3=C1C=CC=C3)NC4(CCCNC4)C(O)=O.Cl

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.