Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-N-(Methanesulfonamido)phenylboronic acid, 95%, Thermo Scientific™

Catalog No. AC454250050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454250050 5 g
AC454250010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454250050 Supplier Thermo Scientific Chemicals Supplier No. 454250050

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 148355-75-3
Molecular Formula C7H10BNO4S
Molecular Weight (g/mol) 215.04
InChI Key XUIQQIRLFMCWLN-UHFFFAOYSA-N
Synonym 3-methylsulfonylamino phenylboronic acid, 3-methylsulfonamido phenylboronic acid, 3-methanesulfonylaminophenylboronic acid, 3-methylsulfonylaminophenylboronic acid, 3-methanesulfonamidophenyl boronic acid, 3-methanesulfonamido phenyl boronic acid, 3-methanesulfonamidophenylboronic acid, 3-methylsulphonylamino benzeneboronic acid, n-3-methanesulfonamidephenylboronic acid, 3-methylsulfonylamino benzeneboronic acid
PubChem CID 2773535
IUPAC Name [3-(methanesulfonamido)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)NS(=O)(=O)C)(O)O

Specifications

Melting Point 255.0°C to 258.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 215.04
Percent Purity 95%
Chemical Name or Material 3-N-(Methanesulfonamido)phenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.