Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(N-tert-Butoxycarbonyl-N-methylamino)pyrrolidine 96.0+%, TCI America™
SDP

Catalog No. M10965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10965G 5 g
M109625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10965G Supplier TCI America Supplier No. M10965G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 172478-00-1
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD00191665
InChI Key XYKYUXYNQDXZTD-UHFFFAOYSA-N
Synonym 3-(N-Boc-N-methylamino)pyrrolidine
PubChem CID 2761404
IUPAC Name tert-butyl N-methyl-N-pyrrolidin-3-ylcarbamate
SMILES CC(C)(C)OC(=O)N(C)C1CCNC1

Specifications

Boiling Point 96°C
Color Colorless
Quantity 5 g
Formula Weight 200.28
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(N-tert-Butoxycarbonyl-N-methylamino)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.