Learn More
Medchemexpress LLC 3-(naphthalen-1-yloxy)propane-1,2-diol | 36112-95-5 | MFCD00673941 | 99.3% | 218.25 g/mol | C13H14O3 | 100 MG
SDP
Catalog No. 5000212720
Encompass_Preferred
Propranolol glycol is an endogenous metabolite of propranolol reported to exhibit instantaneous anticonvulsant activity. It is provided as a white to off-white solid for research use and is identified by CAS 36112-95-5, molecular formula C13H14O3, and molecular weight 218.25 g/mol.
  • Purity 99.3% suitable for analytical and biological assays.
  • Solid form; white to off-white appearance for easy handling.
  • Stable under recommended storage for long-term retention.
  • Relevant for metabolic and pharmacological studies of propranolol.
  • Supplied in a 100 MG quantity for small-scale research applications.
Catalog No. 50-002-12720 Supplier Medchemexpress LLC Supplier No. HY101240100MG
Propranolol glycol is an endogenous metabolite of propranolol reported to exhibit instantaneous anticonvulsant activity. It is provided as a white to off-white solid for research use and is identified by CAS 36112-95-5, molecular formula C13H14O3, and molecular weight 218.25 g/mol.
  • Purity 99.3% suitable for analytical and biological assays.
  • Solid form; white to off-white appearance for easy handling.
  • Stable under recommended storage for long-term retention.
  • Relevant for metabolic and pharmacological studies of propranolol.
  • Supplied in a 100 MG quantity for small-scale research applications.