Learn More
Medchemexpress LLC 3-NBD-C12 cholesterol | 186833-02-3 | MFCD00075336 | 10 MG
SDP
Catalog No. 5000297460
Encompass_Preferred
3-NBD-C12 cholesterol is an NBD-tagged fluorescent cholesterol analog used to study cholesterol uptake, membrane incorporation, intracellular trafficking, and metabolism in biological systems. The NBD fluorophore enables visible-range fluorescence detection for microscopy and spectroscopy; supplied as a solid for research use.
  • NBD fluorophore with typical excitation ≈465 nm and emission ≈535 nm
  • Suitable for studies of membrane dynamics and cholesterol trafficking
  • Soluble in DMF and DMSO for stock solutions with recommended handling
  • Solid, yellow to orange appearance for easy visual identification
  • Store protected from light; use cold storage for solutions to maintain stability
Catalog No. 50-002-97460 Supplier Medchemexpress LLC Supplier No. HYD162110MG
3-NBD-C12 cholesterol is an NBD-tagged fluorescent cholesterol analog used to study cholesterol uptake, membrane incorporation, intracellular trafficking, and metabolism in biological systems. The NBD fluorophore enables visible-range fluorescence detection for microscopy and spectroscopy; supplied as a solid for research use.
  • NBD fluorophore with typical excitation ≈465 nm and emission ≈535 nm
  • Suitable for studies of membrane dynamics and cholesterol trafficking
  • Soluble in DMF and DMSO for stock solutions with recommended handling
  • Solid, yellow to orange appearance for easy visual identification
  • Store protected from light; use cold storage for solutions to maintain stability