Learn More
Medchemexpress LLC 3-nitro-2H-1-benzopyran-2-one | 28448-04-6 | 100.0% | 191.14 | 100 MG
SDP
Catalog No. 5000439154
Encompass_Preferred
3-Nitrocoumarin is an inhibitor for phospholipase C (PLC) that blocks the generation of inositol triphosphate (IP3), thereby affecting the calcium signaling pathway. It is intended for research use only.
  • Inhibits phospholipase C.
  • Blocks the generation of inositol triphosphate (IP3).
  • Affects the calcium signaling pathway.
  • Inhibits glucose-induced increase of intracellular calcium ions in yeast strain WPY260.
  • Inhibits PLC in Neurospora crassa with an IC50 of 10 μg/mL.
  • Inhibits the growth of N. crassa and causes morphological changes of apical hyperbranching in hyphae.
Catalog No. 50-004-39154 Supplier Medchemexpress LLC Supplier No. HY111919100MG
3-Nitrocoumarin is an inhibitor for phospholipase C (PLC) that blocks the generation of inositol triphosphate (IP3), thereby affecting the calcium signaling pathway. It is intended for research use only.
  • Inhibits phospholipase C.
  • Blocks the generation of inositol triphosphate (IP3).
  • Affects the calcium signaling pathway.
  • Inhibits glucose-induced increase of intracellular calcium ions in yeast strain WPY260.
  • Inhibits PLC in Neurospora crassa with an IC50 of 10 μg/mL.
  • Inhibits the growth of N. crassa and causes morphological changes of apical hyperbranching in hyphae.