Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitro-4-(1-pyrrolidinyl)benzaldehyde, 97%, Thermo Scientific™

Catalog No. AAB2236403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2236403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2236403 Supplier Thermo Scientific Chemicals Supplier No. B2236403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 284679-97-6
Molecular Formula C11H12N2O3
Molecular Weight (g/mol) 220.228
MDL Number MFCD00455104
InChI Key AONHBESCCBXPAD-UHFFFAOYSA-N
PubChem CID 599518
IUPAC Name 3-nitro-4-pyrrolidin-1-ylbenzaldehyde
SMILES C1CCN(C1)C2=C(C=C(C=C2)C=O)[N+](=O)[O-]

Specifications

Melting Point 102°C to 104°C
Quantity 1 g
Sensitivity Air Sensitive
Synonym 3-nitro-4-pyrrolidin-1-yl benzaldehyde, 3-nitro-4-1-pyrrolidino benzaldehyde, 3-nitro-4-pyrrolidin-1-yl-benzaldehyde, 3-nitro-4-1-pyrrolidinyl benzaldehyde, 3-nitro-4-pyrrolidinylbenzaldehyde, 3-nitro-4-1-pyrrolidinyl benzenecarbaldehyde, 3-nitro-4-pyrrolidino-benzaldehyde, 3-nitro-4-1-pyrrolidinyl benzaldehyde #, benzaldehyde,3-nitro-4-1-pyrrolidinyl, benzaldehyde, 3-nitro-4-1-pyrrolidinyl
Formula Weight 220.23
Percent Purity 97%
Chemical Name or Material 3-Nitro-4-(1-pyrrolidinyl)benzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.