Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Nitro-4-dimethylaminoazobenzene 97.0+%, TCI America™
SDP

Catalog No. N019125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N019125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N019125G Supplier TCI America Supplier No. N019125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3837-55-6
Molecular Formula C14H14N4O2
Molecular Weight (g/mol) 270.292
MDL Number MFCD00059825
InChI Key BISWHFCOHYEFQW-UHFFFAOYSA-N
Synonym 4-Dimethylamino-3′C-nitroazobenzene
PubChem CID 19690
IUPAC Name N,N-dimethyl-4-[(3-nitrophenyl)diazenyl]aniline
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC(=CC=C2)[N+](=O)[O-]

Specifications

Melting Point 161°C
Quantity 25 g
Formula Weight 270.29
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Nitro-4-dimethylaminoazobenzene
RTECSNumber BX7050000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.