Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitro-4-quinolinol 98.0+%, TCI America™
SDP

Catalog No. N0992200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0992200MG 200 mg
N09921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0992200MG Supplier TCI America Supplier No. N0992200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50332-66-6
Molecular Formula C9H6N2O3
Molecular Weight (g/mol) 190.158
MDL Number MFCD00277722
InChI Key ZWISCKSGNCMAQO-UHFFFAOYSA-N
Synonym 4-Hydroxy-3-nitroquinoline
PubChem CID 316988
IUPAC Name 3-nitro-1H-quinolin-4-one
SMILES C1=CC=C2C(=C1)C(=O)C(=CN2)[N+](=O)[O-]

Specifications

Melting Point 361°C
Color Yellow
Quantity 200 mg
Formula Weight 190.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitro-4-quinolinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.