Learn More
Medchemexpress LLC HY-113248 100mg Medchemexpress, 3-Nitro-L-tyrosine CAS:621-44-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY113248100MG
Encompass_Preferred
Medchemexpress, HY-113248 100mg 3-Nitro-L-tyrosine CAS:621-44-3 3-Nitro-L-tyrosine is a biomarker of nitrogen free radical species modified proteins in systemic autoimmunogenic conditions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0849