Learn More
Medchemexpress LLC 3-nitro-N-(4-phenoxyphenyl) benzamide | 316146-57-3 | MFCD01596399, MFCD32201095 | 99.3% | 334.33 g·mol⁻1 | C19H14N2O4 | 5 MG
SDP
Catalog No. 5000264673
Encompass_Preferred
ICA-105574 is a small-molecule hERG channel activator used in cardiac electrophysiology research. It potentiates hERG currents primarily by removing channel inactivation and is used to probe mechanisms of delayed repolarization and arrhythmia.
  • Potent hERG channel activator that removes channel inactivation.
  • Used to study cardiac repolarization and arrhythmia mechanisms.
  • Off-white to light yellow solid with high purity (99.3%).
  • Molecular formula C19H14N2O4; molecular weight 334.33 g·mol⁻1.
  • Highly soluble in DMSO (≈250 mg/mL); ultrasonic assistance recommended.
  • Powder stable at -20°C for up to 3 years; solvent storage limits apply.
  • Supplied in small research quantities suitable for in vitro studies.
Catalog No. 50-002-64673 Supplier Medchemexpress LLC Supplier No. HY1247025MG
ICA-105574 is a small-molecule hERG channel activator used in cardiac electrophysiology research. It potentiates hERG currents primarily by removing channel inactivation and is used to probe mechanisms of delayed repolarization and arrhythmia.
  • Potent hERG channel activator that removes channel inactivation.
  • Used to study cardiac repolarization and arrhythmia mechanisms.
  • Off-white to light yellow solid with high purity (99.3%).
  • Molecular formula C19H14N2O4; molecular weight 334.33 g·mol⁻1.
  • Highly soluble in DMSO (≈250 mg/mL); ultrasonic assistance recommended.
  • Powder stable at -20°C for up to 3 years; solvent storage limits apply.
  • Supplied in small research quantities suitable for in vitro studies.