Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Nitroacetophenone 98.0+%, TCI America™
SDP

Catalog No. N0109100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0109100G 100 g
N010925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0109100G Supplier TCI America Supplier No. N0109100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121-89-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD00007259
InChI Key ARKIFHPFTHVKDT-UHFFFAOYSA-N
Synonym 3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
PubChem CID 8494
IUPAC Name 1-(3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 78°C
Boiling Point 202°C
Quantity 100 g
Formula Weight 165.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Nitroacetophenone
EINECSNumber (3)-1243
RTECSNumber AM9625000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.