Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitroaniline, 98%

Catalog No. p-7080897
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1062922 100 g
AAA1062936 500 g
AAA106290E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1062922 Supplier Thermo Scientific Chemicals Supplier No. A1062922
Only null left
Add to Cart

CAS: 99-09-2 | C6H6N2O2 | 138.13 g/mol

3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.

Solubility
It is soluble in water 0.1 g/100 ml (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, chloroformates, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-09-2
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007782
InChI Key XJCVRTZCHMZPBD-UHFFFAOYSA-N
Synonym m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine
PubChem CID 7423
IUPAC Name 3-nitroaniline
SMILES NC1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point 112°C to 114°C
Density 0.901
Boiling Point 306°C
Quantity 100 g
UN Number UN1661
Beilstein 636962
Merck Index 14,6582
Solubility Information It is soluble in water 0.1g/100 ml (20°C).
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 3-Nitroaniline
Show More Show Less
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H310-H330-H373-H302
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement P260-P262-P264b-P270-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROANILINES
EINECSNumber 202-729-1
RTECSNumber BY6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.