Learn More
Medchemexpress LLC 3-nitrobenzaldehyde | 99-61-6 | MFCD00007249 | 99.6% | 151.12 g/mol | C7H5NO3 | 100 G
SDP
Catalog No. 5000300180
Encompass_Preferred
3-Nitrobenzaldehyde is an aromatic aldehyde used as a synthetic building block and biochemical reagent for organic synthesis and research. It functions as an intermediate in the preparation of pharmaceuticals, dyes, flavors, and other fine chemicals, and is supplied with documented purity, solubility, and storage guidance for laboratory use.
  • Used as a synthetic intermediate in organic synthesis.
  • Applicable to the synthesis of pharmaceuticals, dyes, and flavor compounds.
  • Available with documented high purity for research applications.
  • Soluble in DMSO at approximately 200 mg/mL (may require sonication).
  • Stable when stored under nitrogen at room temperature; long-term storage in solvent at -80°C or -20°C.
Catalog No. 50-003-00180 Supplier Medchemexpress LLC Supplier No. HYW017410100G
3-Nitrobenzaldehyde is an aromatic aldehyde used as a synthetic building block and biochemical reagent for organic synthesis and research. It functions as an intermediate in the preparation of pharmaceuticals, dyes, flavors, and other fine chemicals, and is supplied with documented purity, solubility, and storage guidance for laboratory use.
  • Used as a synthetic intermediate in organic synthesis.
  • Applicable to the synthesis of pharmaceuticals, dyes, and flavor compounds.
  • Available with documented high purity for research applications.
  • Soluble in DMSO at approximately 200 mg/mL (may require sonication).
  • Stable when stored under nitrogen at room temperature; long-term storage in solvent at -80°C or -20°C.