Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzenesulfonamide, 98%, Thermo Scientific™

Catalog No. AC292620100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC292620100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC292620100 Supplier Thermo Scientific Chemicals Supplier No. 292620100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-52-8
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.184
MDL Number MFCD00007935
InChI Key TXTQURPQLVHJRE-UHFFFAOYSA-N
Synonym benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid
PubChem CID 67138
IUPAC Name 3-nitrobenzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 164°C to 168°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula O2NC6H4SO2NH2
Quantity 10 g
Beilstein 11, 70
Formula Weight 202.18
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 3-Nitrobenzenesulfonamide, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-477-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.