Learn More
Medchemexpress LLC 3-nitrobenzoic acid | 121-92-6 | MFCD00007251 | 99.2% | 167.12 | C7H5NO4 | 100g
SDP
Catalog No. 5000266596
Encompass_Preferred
3-Nitrobenzoic acid (m-Carboxynitrobenzene m-Nitrobenzenecarboxylic acid m-Nitrobenzoic acid) is an antioxidant and antibacterial agent that can kill bacteria and fungi 3-Nitrobenzoic acid can be degraded or reduced by certain bacteria (such as Pseudomonas) and fungi (such as white rot fungi) into aldehydes and alcohols[1][2]
Catalog No. 50-002-66596 Supplier Medchemexpress LLC Supplier No. HYY1313100G
3-Nitrobenzoic acid (m-Carboxynitrobenzene m-Nitrobenzenecarboxylic acid m-Nitrobenzoic acid) is an antioxidant and antibacterial agent that can kill bacteria and fungi 3-Nitrobenzoic acid can be degraded or reduced by certain bacteria (such as Pseudomonas) and fungi (such as white rot fungi) into aldehydes and alcohols[1][2]