Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzotrifluoride, 97%

Catalog No. AAL0594822
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0594822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0594822 Supplier Thermo Scientific Chemicals Supplier No. L0594822
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 98-46-4 | C7H4F3NO2 | 191.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-46-4
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD00007260
InChI Key WHNAMGUAXHGCHH-UHFFFAOYSA-N
Synonym 3-nitrobenzotrifluoride, 1-nitro-3-trifluoromethyl benzene, m-nitrobenzotrifluoride, 3-trifluoromethyl nitrobenzene, m-nitrotrifluorotoluene, benzene, 1-nitro-3-trifluoromethyl, 1,3-nitrobenzotrifluoride, m-nitrotrifluortoluol, usaf ma-5, 3-trifluoromethylnitrobenzene
PubChem CID 7386
IUPAC Name 1-nitro-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(F)(F)F

Specifications

Melting Point -5°C
Density 1.435
Boiling Point 201°C to 205°C
Flash Point 87°C (188°F)
Odor Characteristic
Refractive Index 1.472
Quantity 100 g
UN Number UN2306
Beilstein 2051362
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 3-Nitrobenzotrifluoride
Show More Show Less
Hazard Category H227-H302+H312-H315-H319-H330-H335
Hazard Statement GHS H Statement
H301-H311-H330-H227-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROBENZOTRIFLUORIDES, LIQUID
EINECSNumber 202-670-1
RTECSNumber XT3500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.