Learn More
Medchemexpress LLC HY-111919 10mg Medchemexpress, 3-Nitrocoumarin CAS:28448-04-6 Purity:>98%
SDP
Catalog No. 501968983
Encompass_Preferred
Medchemexpress, HY-111919 10mg 3-Nitrocoumarin CAS:28448-04-6 3-Nitrocoumarin (3-NC) is a potent and selective Phospholipase C-γ (PLC-γ) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8983 Supplier Medchemexpress LLC Supplier No. HY11191910MG
Medchemexpress, HY-111919 10mg 3-Nitrocoumarin CAS:28448-04-6 3-Nitrocoumarin (3-NC) is a potent and selective Phospholipase C-γ (PLC-γ) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.