Learn More
TARGETMOL CHEMICALS INC 3-Nitrocoumarin 25MG
SDP
Catalog No. 502880651
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 3-Nitrocoumarin (3-nitrochromen-2-one) (3-NC) is a potent and selective Phospholipase C-gamma (PLCgamma) inhibitor. Purity 97.28%
Catalog No. 50-288-0651 Supplier TARGETMOL CHEMICALS INC Supplier No. T1011525MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 3-Nitrocoumarin (3-nitrochromen-2-one) (3-NC) is a potent and selective Phospholipase C-gamma (PLCgamma) inhibitor. Purity 97.28%