Learn More
Apexbio Technology LLC 3-Nitrocoumarin 28448-04-6 5mg
SDP
Catalog No. 5000097170
Encompass_Preferred
3-Nitrocoumarin (CAS 28448-04-6) is a selective inhibitor of phospholipase C- (PLC- ) an enzyme involved in cellular signal transduction through the hydrolysis of membrane phospholipids and subsequent generation of secondary messengers By suppressing PLC- catalytic activity 3-Nitrocoumarin disrupts related intracellular signaling pathways thereby enabling investigation of PLC- -mediated mechanisms in various physiological and pathological contexts This compound is widely utilized in studies focused on elucidating the roles of PLC- in disease models and fundamental biomedical research
Catalog No. 50-000-97170 Supplier Apexbio Technology LLC Supplier No. A46395
3-Nitrocoumarin (CAS 28448-04-6) is a selective inhibitor of phospholipase C- (PLC- ) an enzyme involved in cellular signal transduction through the hydrolysis of membrane phospholipids and subsequent generation of secondary messengers By suppressing PLC- catalytic activity 3-Nitrocoumarin disrupts related intracellular signaling pathways thereby enabling investigation of PLC- -mediated mechanisms in various physiological and pathological contexts This compound is widely utilized in studies focused on elucidating the roles of PLC- in disease models and fundamental biomedical research