Learn More
Medchemexpress LLC 3-nitrooxypropanol | 100502-66-7 | MFCD31731012 | 95.0% | C3H7NO4 | 10MG
SDP
Catalog No. 5000261257
Encompass_Preferred
Abrucomstat (3-nitrooxypropanol) is a small-molecule inhibitor of methyl coenzyme M reductase that reduces ruminal methanogenesis. It is used in research on enteric methane mitigation and animal nutrition. Reported properties include CAS 100502-66-7, molecular formula C3H7NO4, molecular weight 121.09, and reported purity 95.0%.
  • Inhibits methyl coenzyme M reductase and reduces methane production in ruminants.
  • Orally active mechanism suited for in vivo rumen studies.
  • Supports research on enteric methane mitigation and feed efficiency.
  • Small molecular weight with a simple chemical structure for analytical work.
  • Supplied with a certificate of analysis indicating reported purity.
Catalog No. 50-002-61257 Supplier Medchemexpress LLC Supplier No. HY13956610MG
Abrucomstat (3-nitrooxypropanol) is a small-molecule inhibitor of methyl coenzyme M reductase that reduces ruminal methanogenesis. It is used in research on enteric methane mitigation and animal nutrition. Reported properties include CAS 100502-66-7, molecular formula C3H7NO4, molecular weight 121.09, and reported purity 95.0%.
  • Inhibits methyl coenzyme M reductase and reduces methane production in ruminants.
  • Orally active mechanism suited for in vivo rumen studies.
  • Supports research on enteric methane mitigation and feed efficiency.
  • Small molecular weight with a simple chemical structure for analytical work.
  • Supplied with a certificate of analysis indicating reported purity.