Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrophenetole 99.0+%, TCI America™
SDP

Catalog No. N021525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N021525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N021525G Supplier TCI America Supplier No. N021525G

Chemical Identifiers

CAS 621-52-3
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00024472
InChI Key LFOLBPDHVGDKGJ-UHFFFAOYSA-N
Synonym 1-Ethoxy-3-nitrobenzene
PubChem CID 69308
IUPAC Name 1-ethoxy-3-nitrobenzene
SMILES CCOC1=CC=CC(=C1)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 167.16
Percent Purity ≥99.0% (GC)
Physical Form Crystals
Chemical Name or Material 3-Nitrophenetole
EINECSNumber (3)-0783
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.