Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrophenoxyacetic Acid 98.0+%, TCI America™
SDP

Catalog No. N02245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02245G 5 g
N022425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02245G Supplier TCI America Supplier No. N02245G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1878-88-2
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00059826
InChI Key BNRRQAASFDGMMQ-UHFFFAOYSA-N
Synonym 3-nitrophenoxyacetic acid, 3-nitrophenoxy acetic acid, 2-3-nitrophenoxy acetic acid, m-nitrophenoxyacetic acid, 3-nitrophenoxy-acetic acid, acmc-209eqk, 3-nitrophenoxyaceticacid, 3-nitro-phenoxy-acetic acid
PubChem CID 74656
SMILES C1=CC(=CC(=C1)OCC(=O)O)[N+](=O)[O-]

Specifications

Melting Point 153°C
Color Yellow
Quantity 5 g
Formula Weight 197.15
Percent Purity ≥98.0% (T,GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophenoxyacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.