Learn More
Medchemexpress LLC 3-Nitrophenyl disulfide | 537-91-7 | 99.2% | 308.33 | 25 MG
SDP
Catalog No. 5000366734
Encompass_Preferred
3-Nitrophenyl disulfide (Bis(3-Nitrophenyl) disulfide) is an inhibitor of mannitol-1-phosphate dehydrogenase (M1PDH) with an IC50 of 3 μM. It also demonstrates antiparasitic activity.
  • Inhibits mannitol-1-phosphate dehydrogenase (M1PDH)
  • Demonstrates antiparasitic activity
Catalog No. 50-003-66734 Supplier Medchemexpress LLC Supplier No. 25MGHYW009213
3-Nitrophenyl disulfide (Bis(3-Nitrophenyl) disulfide) is an inhibitor of mannitol-1-phosphate dehydrogenase (M1PDH) with an IC50 of 3 μM. It also demonstrates antiparasitic activity.
  • Inhibits mannitol-1-phosphate dehydrogenase (M1PDH)
  • Demonstrates antiparasitic activity