Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Nitrophenyl disulfide | 537-91-7 | 99.23% | 308.33 | 1 ML
SDP

Catalog No. 5000451508
Encompass_Preferred

3-Nitrophenyl disulfide (Bis(3-Nitrophenyl) disulfide) is an inhibitor of mannitol-1-phosphate dehydrogenase (M1PDH) with an IC50 of 3 μM. It also demonstrates antiparasitic activity.

  • Inhibitor of mannitol-1-phosphate dehydrogenase (M1PDH)
  • Exhibits antiparasitic activity
  • IC50 of 3 μM for M1PDH
  • Solid appearance, light yellow to green yellow color
  • Molecular formula: C12H8N2O4S2
  • Molecular weight: 308.33
  • Solubility in DMSO: 100 mg/mL
  • Storage: -20°C for 3 years (powder), 4°C for 2 years (powder); -80°C for 6 months (in solvent), -20°C for 1 month (in solvent)

Catalog No. 50-004-51508 Supplier Medchemexpress LLC Supplier No. HYW00921310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Nitrophenyl disulfide (Bis(3-Nitrophenyl) disulfide) is an inhibitor of mannitol-1-phosphate dehydrogenase (M1PDH) with an IC50 of 3 μM. It also demonstrates antiparasitic activity.

  • Inhibitor of mannitol-1-phosphate dehydrogenase (M1PDH)
  • Exhibits antiparasitic activity
  • IC50 of 3 μM for M1PDH
  • Solid appearance, light yellow to green yellow color
  • Molecular formula: C12H8N2O4S2
  • Molecular weight: 308.33
  • Solubility in DMSO: 100 mg/mL
  • Storage: -20°C for 3 years (powder), 4°C for 2 years (powder); -80°C for 6 months (in solvent), -20°C for 1 month (in solvent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.